missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD47 Monoclonal Antibody (2D3), NovaFluor™ Yellow 730, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD47 |
|---|---|
| Clone | 2D3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30054856
|
Invitrogen™
H065T03Y07-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054900
|
Invitrogen™
H065T02Y07-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow CytometrySpecifications
| CD47 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q08722 | |
| 961 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 2D3 | |
| Flow Cytometry | |
| NovaFluor Yellow 730 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD47 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title