Learn More
Invitrogen™ Ongericimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559618
 View more versions of this product
Product Code. 30284158
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ongericimab, also known as a recombinant humanized anti-PCSK9 monoclonal antibody, is primarily used to treat hypercholesterolemia by reducing low-density lipoprotein cholesterol (LDL-C) levels. The PCSK9 gene, located on chromosome 1p32.3, spans 22 kilobases and consists of 12 exons and 11 introns. It encodes a 692 amino acid glycoprotein involved in lipid metabolism, comprising a signal peptide (amino acids 1-30), a prosegment or inhibitory prodomain (amino acids 31-152), a subtilisin-like catalytic domain (amino acids 153-452), and a cysteine-histidine-rich C-terminal domain. PCSK9 plays a pivotal role in cholesterol homeostasis by promoting the degradation of low-density lipoprotein receptors (LDLR) in the liver, thereby increasing plasma LDL levels. Mutations in PCSK9 can lead to disorders such as familial hypercholesterolemia. In addition to its well-established role in cardiovascular diseases, emerging research indicates that PCSK9 variants may influence cancer outcomes and that PCSK9 inhibition can enhance the efficacy of immune checkpoint therapies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ongericimab Humanized
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG4SP