missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD48 Monoclonal Antibody (HM48-1), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | CD48 |
|---|---|
| Clone | HM48-1 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30054872
|
Invitrogen™
M031T03B09-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055571
|
Invitrogen™
M031T02B09-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD48 Monoclonal antibody specifically detects CD48 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD48 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| Mouse | |
| P18181 | |
| 12506 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| HM48-1 | |
| Flow Cytometry | |
| NovaFluor Blue 690 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
| IgG | |
| Affinity chromatography | |
| Cd48 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title