missing translation for 'onlineSavingsMsg'
Learn More

CD49f Rat anti-Human, PE, Clone: GOH3, BD

Rat Monoclonal Antibody

71.14 € - 279.74 €

Specifications

Antigen CD49f
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
Host Species Rat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15891439
View Documents
BD Biosciences
561894
25 Tests
ONLINE EXCLUSIVE PRICE
72.80 €
71.14 € / Each
Save 1.66 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15898398
View Documents
BD Biosciences
555736
100 Tests
ONLINE EXCLUSIVE PRICE
292.00 €
279.74 € / Each
Save 12.26 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GoH3 monoclonal antibody specifically binds to CD49f which is also known as integrin α6 chain. CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The integrin α6 subunit associates with the integrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is expressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.

Specifications

Specifications

CD49f
Monoclonal
Rat
Canine, Human, Mouse, Pig
Mouse mammary tumor cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.