missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Antibody (CL12420) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21215JF646
This item is not returnable.
View return policy
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CD5 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| This antibody was developed using a synthetic peptide derived from P06127, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
| 921 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL12420 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction