missing translation for 'onlineSavingsMsg'
Learn More

CD5 Mouse, PerCP, Clone: L17F12, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB14-PCP-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961538

  • 215.00 € / 25 tests
Estimated Shipment: 20-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD5
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD5
Protein A/G Purified
RUO
921
IgG2a κ
Flow Cytometry
L17F12
PerCP
NP_055022
Mouse
25 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers