missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD52 Antibody (SR2268), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21870-100ul
This item is not returnable.
View return policy
Description
CD52 Monoclonal antibody specifically detects CD52 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CD52 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2268 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 | |
| A synthesized peptide derived from human CD52 (Uniprot #: Q64389) | |
| 100 μg | |
| Cancer, Immunology | |
| 1043 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction