missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Rabbit anti-Human, Clone: 161-2 (53/2), Novus Biologicals™
Rabbit Monoclonal Antibody
255.00 € - 612.00 €
Specifications
| Antigen | CD53 |
|---|---|
| Clone | 161-2 (53/2) |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Applications | Flow Cytometry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351942
|
Bio-Techne
NBP3-12077-0.025MG |
0.025 mg |
255.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380002
|
Novus Biologicals
NBP3-12077-0.2MG |
0.2 mg |
612.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
| CD53 | |
| 1 mg/mL | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
| This antibody was raised by immunizng mice with stimulated human leukocytes. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 161-2 (53/2) | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Rabbit | |
| Immunology | |
| PBS | |
| 963 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title