missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Rabbit anti-Human, Cynomolgus Monkey, Clone: 104, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90952-50UL
This item is not returnable.
View return policy
Description
CD53 Monoclonal specifically detects CD53 in Human, Cynomolgus Monkey samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
| CD53 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 104 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
| Recombinant Cynomolgus CD53 Protein | |
| 50 μg | |
| Immunology | |
| 963 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction