missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD54 (ICAM-1) Monoclonal Antibody (YN1/1.7.4), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | CD54 (ICAM-1) |
|---|---|
| Clone | YN1/1.7.4 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056203
|
Invitrogen™
M057T02Y06-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056329
|
Invitrogen™
M057T03Y06-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD54 (ICAM-1) Monoclonal antibody specifically detects CD54 (ICAM-1) in Mouse samples. It is validated for Flow Cytometry
Description: The YN1/1.7.4 monoclonal antibody reacts with mouse CD54 (InterCellular Adhesion Molecule-1, ICAM-1), a 90-110 kDa transmembrane glycoprotein expressed by monocytes, lymphocytes, dendritic cells, and endothelial cells. Expression of CD54 is upregulated on activated lymphocytes. Interaction of CD54 with its ligands CD11a and CD11b is important in the inflammatory response. The YN1/1.7.4 monoclonal antibody recognizes a different epitope than the eBioKAT-1 monclonal antibody. Applications Reported: The YN1/1.7.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: The YN1/1.7.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.4 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Each product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer . NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. This NovaFluor conjugate has been updated to ship with CellBlox Plus Blocking Buffer (Cat. No. (<a href='https://www.thermofisher.com/order/catalog/product/C001T06F01'>C001T06F01</a>)). This buffer contains formulation improvements over CellBlox. CellBlox Plus Blocking Buffer is required for optimal staining with NovaFluor conjugates and should be used in all experiments where NovaFluor conjugates are used. Whenever possible, we recommend adding CellBlox Plus Blocking Buffer to antibody cocktails/master mixes prior to combining with cells. Add 5 μL per sample (regardless of the number of NovaFluors in your panel) to use the antibody cocktail as intended. For single-color controls, use 5 μL of CellBlox Blocking Buffer per 100 μL of cell sample containing 10^3 to 10^8 cells. NovaFluor conjugates are based on Phiton™ technology utilizing novel nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. <a href = https://www.thermofisher.com/us/en/home/life-science/cell-analysis/flow-cytometry/novafluor-dyes.html>Learn more </a> Excitation: 552 nm; Emission: 700 nm; Laser: 561 nm (Yellow) Laser Our internal testing shows that NovaFluor Yellow 700 non-specifically stains B cells in SJL mice. Non-specific staining has not been observed in BALB/c or C57BL/6 mice. Other strains have not been tested. See the Antibody Testing Data for an example of this strain-dependent difference.Specifications
| CD54 (ICAM-1) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| BB2; CD54; cell surface glycoprotein P3.58; Human rhinovirus receptor; ICAM; Icam1; ICAM-1; ICAM-1; CD54 homolog; Intercellular adhesion molecule 1; intercellular adhesion molecule 1 (CD54), human rhinovirus receptor; intercellular adhesion molecule-1; intercellular adhesion molecule-1 precursor; Ly 47; Ly-47; major group rhinovirus receptor; MALA2; MALA-2; MyD10; P3.58; sCD54; soluble CD54; Surface antigen of activated B cells | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| YN1/1.7.4 | |
| Flow Cytometry | |
| NovaFluor Yellow 700 | |
| Rat | |
| Mouse | |
| P13597 | |
| 15894 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| ICAM1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title