missing translation for 'onlineSavingsMsg'
Learn More

CD54 / ICAM-1 Mouse, FITC, Clone: 15.2, enQuireBio™

Product Code. 15922098
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15922098 100 Tests 100 tests
15932098 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15922098 Supplier EnquireBio Supplier No. QAB47F100Tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD54 / ICAM-1
Applications Flow Cytometry
Classification Monoclonal
Clone 15.2
Concentration 5μL/Test
Conjugate FITC
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Gene Accession No. NP_000192
Gene Symbols ICAM1
Host Species Mouse
Purification Method Protein A/G Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Content And Storage Store at 2-4°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.