Learn More
CD54 Mouse anti-Human, APC, Clone: HA58, BD
Mouse Monoclonal Antibody
55.36 € - 314.00 €
Specifications
| Antigen | CD54 |
|---|---|
| Clone | HA58 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15841449
|
BD Biosciences
561899 |
25 Tests |
77.05 € 55.36 € / Each Save 21.69 € 28% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15859768
|
BD Biosciences
559771 |
100 Tests |
314.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.
Specifications
| CD54 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| ICAM-1; intercellular adhesion molecule 1; BB2; P3.58 |
| HA58 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IFN-γ-treated BM314 colonic carcinoma cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.