Learn More
CD55/DAF Antibody (S01-7H4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19600-100ul
 View more versions of this product
Product Code. 18629683
  • 645.00 € / 100µL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD55/DAF
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
S01-7H4
Unconjugated
CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
A synthetic peptide of human CD55/DAF (Uniprot # P08174)
100 μg
Immunology
1604
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.