missing translation for 'onlineSavingsMsg'
Learn More

CD56 / NCAM Mouse, APC, Clone: MY31, enQuireBio™

Mouse Monoclonal Antibody

183.75 € - 367.50 €

Specifications

Antigen CD56 / NCAM
Clone MY31
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15982098
View Documents
EnquireBio
QAB48-APC-25Tests
25 Tests
183.75 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15972098
View Documents
EnquireBio
QAB48-APC-100Tests
100 Tests
367.50 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD56 / NCAM
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
4684
IgG1 κ
Protein A/G Purified
MY31
Flow Cytometry
APC
RUO
NP_000606
NCAM1
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado