missing translation for 'onlineSavingsMsg'
Learn More

CD56 / NCAM1 / NKH1 Mouse, Clone: NCAM1/1496, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 4684-MSM6-P1

 View more versions of this product

Product Code. 15900568

  • ONLINE EXCLUSIVE PRICE
    554.40 €
    478.00 € / 100µg
    Save 76.40 €
    13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD56 / NCAM1 / NKH1
Cell surface
NCAM1/1496
Unconjugated
NP_000606
Mouse
180, 145 and 125kDa
Protein A/G Purified
RUO
4684
IgG2a κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
NCAM1
Recombinant human NCAM1 protein
Cerebellum, Pancreas, Neuroblastoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.