missing translation for 'onlineSavingsMsg'
Learn More

CD57 (Natural Killer Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.

Brand:  Epredia MS-136-R7

 View more versions of this product

Product Code. 12609676

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD57 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Host Species: Mouse

Clone: NK1

Isotype: IgM/κ

Species Reactivity: Human. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: Human peripheral blood mononuclear cells

Molecular Weight: 110 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD57 (Natural Killer Cell Marker) Ab-1
Monoclonal
Unconjugated
Human peripheral blood mononuclear cells
IVD
Primary
Prediluted
Flow Cytometry, Immunohistochemistry (Paraffin)
NK1
Mouse
7 mL
Cell Adhesion
Human
IgM κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.