missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD58/LFA-3 Antibody (TS2/9) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
218.00 € - 529.00 €
Specifications
| Antigen | CD58/LFA-3 |
|---|---|
| Clone | TS2/9 |
| Concentration | 0.98 mg/mL |
| Dilution | Western Blot 1:100 - 1:500, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Neutralization |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18415262
|
Novus Biologicals
NBP2-22542-50ug |
50 ug |
218.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18019623
|
Novus Biologicals
NBP2-22542 |
200 μg |
529.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.Specifications
| CD58/LFA-3 | |
| 0.98 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, Neutralization | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology | |
| P19256 | |
| 965 | |
| Human Leukocyte Function Antigen-3 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| TS2/9 | |
| Western Blot 1:100 - 1:500, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Neutralization | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
| CD58 | |
| IgG1 | |
| Protein A purified | |
| This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title