missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD61 (Integrin beta 3) Monoclonal Antibody (2C9.G3), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
185.00 € - 348.00 €
Specifications
| Antigen | CD61 (Integrin beta 3) |
|---|---|
| Clone | 2C9.G3 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056147
|
Invitrogen™
M053T03B06-A |
100 μg |
348.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055919
|
Invitrogen™
M053T02B06-A |
25 μg |
185.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD61 (Integrin beta 3) Monoclonal antibody specifically detects CD61 (Integrin beta 3) in Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| CD61 (Integrin beta 3) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| Mouse, Rat | |
| O54890 | |
| 16416, 29302 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 2C9.G3 | |
| Flow Cytometry | |
| NovaFluor Blue 610-70S | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
| IgG | |
| Affinity chromatography | |
| ITGB3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title