missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD62L (L-Selectin) Monoclonal Antibody (DREG-56 (DREG56)), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H009T03B01-A
This item is not returnable.
View return policy
Description
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Specifications
| CD62L (L-Selectin) | |
| Monoclonal | |
| 0.2 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P14151 | |
| SELL | |
| Affinity chromatography | |
| RUO | |
| 6402 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG1 κ |
| Flow Cytometry | |
| DREG-56 (DREG56) | |
| NovaFluor Blue 510 | |
| SELL | |
| A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction