missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L / L-Selectin Rat, APC, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
131.25 € - 275.10 €
Specifications
| Antigen | CD62L / L-Selectin |
|---|---|
| Clone | MEL-14 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15972108
|
EnquireBio
QAB49-APC-25ug |
25 μg |
131.25 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15962108
|
EnquireBio
QAB49-APC-100ug |
100 μg |
275.10 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD62L / L-Selectin | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
| 20343 | |
| IgG2a κ | |
| Protein A/G Purified |
| MEL-14 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| NP_001157531 | |
| Sell | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title