Learn More
CD62L / L-Selectin Rat, FITC, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB49-F-100ug
 View more versions of this product
Product Code. 15922118
  • 169.05 € / 100µg
Specifications
CD62L / L-Selectin
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Sell
Protein A/G Purified
RUO
20343
IgG2a κ
Flow Cytometry
MEL-14
FITC
NP_001157531
Rat
100 μg
Primary
Store at 2-4°C.