missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (S06-4G3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20026-100ul
This item is not returnable.
View return policy
Description
CD63 Monoclonal antibody specifically detects CD63 in Mouse samples. It is validated for Western Blot
Specifications
| CD63 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| S06-4G3 | |
| Unconjugated | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| A synthetic peptide of mouse CD63 (Uniprot # P41731) | |
| 100 μg | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction