missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD63 Immunobeads for Exosome Isolation

Product Code. 18629520 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18629520 10 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18629520 Supplier Novus Biologicals™ Supplier No. NBP311769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications

TRUSTED_SUSTAINABILITY

Specifications

Synonym EV isolation, exosome isolation, extracellular vesicle isolation
Content And Storage Store at 4°C.
Target Species Human
Quantity 10 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.