Learn More
CD64 Mouse anti-Human, PE, Clone: 10.1, BD
Mouse Monoclonal Antibody
72.80 € - 239.00 €
Specifications
| Antigen | CD64 |
|---|---|
| Clone | 10.1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811479
|
BD Biosciences
561926 |
25 Tests |
72.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15869508
|
BD Biosciences
558592 |
100 Tests |
239.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 10.1 monoclonal antibody specifically binds to CD64, a 72 kDa type I transmembrane glycoprotein that is a high affinity receptor for human IgG (FcγRI), especially the IgG1 and IgG3 subclasses. CD64 is expressed on monocytes, macrophages, dendritic cells, granulocytes activated with interferon-gamma and early myeloid lineage cells. CD64 associates with a signaling FcRγhomodimer to form the functional high affinity FcγRI complex. CD64 functions in both innate and adaptive immune responses and mediates endocytosis, phagocytosis, antigen presentation, antibody-dependent cellular toxicity, cytokine release and superoxide generation.
Specifications
| CD64 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 10.1 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.