missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEA15 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149032
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
Specifications
| PEA15 | |
| Polyclonal | |
| Unconjugated | |
| PEA15 | |
| 15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 18611, 8682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q15121, Q62048 | |
| PEA15, Pea15a | |
| E. coli-derived recombinant human PEA-15, Ala2-Ala130. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction