missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (3F7D3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 € - 590.00 €
Specifications
| Antigen | CD68/SR-D1 |
|---|---|
| Clone | 3F7D3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18181788
|
Novus Biologicals
NBP2-37265 |
0.1 mL |
590.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668885
|
Novus Biologicals
NBP2-37265-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF.Specifications
| CD68/SR-D1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| CD68 | |
| IgG1 | |
| Affinity Purified |
| 3F7D3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| PBS with 0.05% Sodium Azide | |
| 968 | |
| This CD68/SR-D1 Antibody (3F7D3) was developed against purified recombinant fragment of human CD68 (AA: 42-155) expressed in E. Coli. | |
| Primary | |
| 37.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title