Learn More
CD68/SR-D1 Antibody (ED1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand:  Novus Biologicals NB600-985-0.025MG
 View more versions of this product
Product Code. 18221334
  • 225.00 € / 25µg
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human, Mouse, Rat, Bovine, Feline, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD68/SR-D1
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500RadioImmunoassay 1:100-1:2000, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000
Q4FZY1
CD68
Rat spleen cells
0.025 mg
Cell Biology, Cytokine Research, Immunology
968
Primate, Feline, Bovine, Mouse, Human, Rat, Monkey
Purified
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
ED1
Unconjugated
Phosphate buffered saline. with 0.09% Sodium Azide
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Mouse
Protein A or G purified
RUO
Primary
NB600-985 recognizes a single chain glycoprotein of 110kD that is expressed predominantly on the lysosomal membrane of myeloid cells. Weak cell surface expression also occurs. The antigen is expressed by the majority of tissue macrophages and weakly by peripheral blood granulocytes. Studies have shown that the antigen recognized by ED1 is the rat homologue of human CD68.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1