Learn More
CD7 Antibody (15), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89748PE
Product Code. 18642561
  • 618.00 € / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
CD7 Monoclonal antibody specifically detects CD7 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD7
Monoclonal
PE
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Mouse (Negative), Rat (Negative)
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
15
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Capture
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD7 (Accession#: NP_006128.1; Met 1-Pro 180).
0.1 mL
Cytokine Research, Signal Transduction
924
Store at 4C in the dark.