missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

178.00 € - 392.00 €

Specifications

Antigen CD7
Clone eBio124-1D1 (124-1D1)
Concentration 0.8 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30056250
View Documents
Invitrogen™
H018T03Y03-A
100 Tests
392.00 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30054845
View Documents
Invitrogen™
H018T02Y03-A
25 Tests
178.00 €
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD7
0.8 μg/Test
Monoclonal
Mouse
Human
P09564
924
Primary
4°C, store in dark, DO NOT FREEZE!
eBio124-1D1 (124-1D1)
Flow Cytometry
NovaFluor Yellow 610
RUO
PBS with 0.09% sodium azide; pH 7.2
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41
IgG1 κ
Affinity chromatography
CD7
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.