missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD70 Monoclonal Antibody (118510)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561876
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
Specifications
| CD70 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O55237 | |
| Cd70 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CD27 Ligand/TNFSF7, Gln47-Pro195. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Neutralization | |
| 118510 | |
| Unconjugated | |
| Cd70 | |
| CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 21948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction