missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10084-2-AP-20UL
This item is not returnable.
View return policy
Description
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregcy, irritable bowel syndrome and in HIV patients.Specifications
| CD71 | |
| Polyclonal | |
| Unconjugated | |
| TFRC | |
| CD71, p90, sTfR, T9, TFR, TFR1, TFRC, TR, Transferrin receptor protein 1, TRFR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7037 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02786 | |
| TFRC | |
| CD71 Fusion Protein Ag0123 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction