missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | CD73 |
|---|---|
| Clone | eBioTY/11.8 (TY/11.8) |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055738
|
Invitrogen™
M032T03R05-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055073
|
Invitrogen™
M032T02R05-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD73 Monoclonal antibody specifically detects CD73 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD73 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q61503 | |
| 23959 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioTY/11.8 (TY/11.8) | |
| Flow Cytometry | |
| NovaFluor Red 725 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 2210401F01Rik; 5 nucleotidase, ecto; 5' nucleotidase, ecto; 5'-NT; 5'-nucleotidase; 5'-nucleotidase ecto; 5'-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; NTE; Purine 5-Prime-Nucleotidase | |
| IgG1 | |
| Affinity chromatography | |
| NT5E |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title