missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody.

202.00 € - 841.60 €

Specifications

Antigen CD79a/mb-1 (B-Cell Marker)
Clone SP18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12653757
View Documents
Epredia™
RM-9118-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12613757
View Documents
Epredia™
RM-9118-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
524.00 €
464.94 € / 500µL
Save 59.06 €
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12633757
View Documents
Epredia™
RM-9118-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
962.00 €
841.60 € / 1mL
Save 120.40 €
12% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12693747
View Documents
Epredia™
RM-9118-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
404.00 €
369.73 € / 7mL
Save 34.27 €
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12623757
View Documents
Epredia™
RM-9118-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

A disulphide-linked heterodimer, consisting of CD79a/mb-1 and CD79b/B29 polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas.

Host Species: Rabbit

Clone: SP18

Species Reactivity: Human. Does not react with mouse, rat, canine, monkey and opossum

Epitope: N-terminus

Immunogen: Synthetic peptide derived from N-terminal region of human CD79a protein

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD79a/mb-1 (B-Cell Marker)
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Synthetic peptide derived from N-terminal region of human CD79a protein
SP18
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.