Learn More
CD79A Rabbit, anti-Human, Clone: 1F24, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82747-4-RR-100UL
Description
The CD79A antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CD79A. This antibody recognizes human antigen. The CD79A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD79A | |
| 1F24 | |
| CD79A, Ig alpha, IGA, MB 1, MB 1 membrane glycoprotein, MB1, Surface IgM associated protein | |
| 100 μL | |
| 973 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |