missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8/CD56 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA112171
This item is not returnable.
View return policy
Description
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD56 (clone C5.9, isotype IgG2b).
CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells.
Specifications
| CD8/CD56 | |
| Cocktail | |
| PE | |
| NCAM1 | |
| antigen recognized by monoclonal antibody 5.1H11; CD56; CD-56; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8B; CD8b molecule; CD8B1; CD8beta; LEU2; Leu-2; Leu2 T-lymphocyte antigen; LY3; LYT3; MAL; MSK39; NCAM; NCAM1; N-CAM-1; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-lymphocyte differentiation antigen T8/Leu-2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4684, 925, 926 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 17D8, C5.9, C5.9 | |
| PBS with 0.2% carrier protein and 0.08% sodium azide | |
| P01732, P10966, P13591 | |
| CD8A, CD8B, NCAM1 | |
| human peripheral blood T lymphocytes and KG1a cell line | |
| 50 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1, IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction