Learn More
CD8 Mouse, PerCP, Clone: SK1, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB18-PCP-500Tests
 View more versions of this product
Product Code. 15951598
  • 2929.50 € / 500 tests
Specifications
CD8
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD8A
Protein A/G Purified
RUO
925
IgG1 κ
Flow Cytometry
SK1
PerCP
NP_741969
Mouse
500 Tests
Primary
Store at 2-4°C.