missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 Armenian Hamster, PE, Clone: 16-10A1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand: EnquireBio QAB50-PE-100ug
This item is not returnable.
View return policy
Specifications
| CD80 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Cd80 | |
| Protein A/G Purified | |
| RUO | |
| 12519 | |
| IgG |
| Flow Cytometry | |
| 16-10A1 | |
| PE | |
| XP_017172347 | |
| Armenian Hamster | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction