missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCOLCE Monoclonal Antibody (261845)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561127
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
Specifications
| PCOLCE | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q61398 | |
| PCOLCE | |
| Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1, Gln25-Ala468. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| 261845 | |
| Unconjugated | |
| PCOLCE | |
| astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer | |
| Rat | |
| Protein A/G | |
| RUO | |
| 18542 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction