missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84/SLAMF5 Antibody (152-1D5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11538F
This item is not returnable.
View return policy
Description
CD84/SLAMF5 Monoclonal antibody specifically detects CD84/SLAMF5 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
| CD84/SLAMF5 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 152-1D5 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5LY9B | |
| Spleen cells of a patient with hairy cell leukemia | |
| 0.1 mL | |
| CD Markers, Immunology, Stem Cells | |
| 8832 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction