missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD85j (ILT2) Monoclonal Antibody (HP-F1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H043T03B06-A
This item is not returnable.
View return policy
Description
CD85j, also known as a member of the leukocyte immunoglobulin-like receptor (LIR) family, is a single transmembrane glycoprotein with a long cytoplasmic domain containing four immunoreceptor tyrosine-based inhibitory motifs (ITIMs). These motifs signal through interactions with SHP-1, transducing negative signals that inhibit immune response stimulation. CD85j is expressed on myeloid cells such as monocytes and dendritic cells, as well as on subsets of lymphoid cells including NK, T, and B cells. The expression of CD85j is correlated with leukemias such as acute lymphoblastic leukemia (ALL) and chronic lymphocytic leukemia (CLL). On CD8+ cells, CD85j expression is associated with effector cell function and plays a significant role in viral infections, including HIV, Epstein-Barr virus, and cytomegalovirus (CMV). CD85j binds to MHC class I molecules, including HLA-G, A, F, B27, E, and F, on antigen-presenting cells. The receptor helps control inflammatory responses and cytotoxicity, focusing the immune response and limiting autoreactivity. CD85j belongs to the subfamily B class of LIR receptors, which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and multiple transcript variants encoding different isoforms have been identified for this gene. Its gene is located in a cluster at chromosomal region 19q13.4.
Specifications
| CD85j (ILT2) | |
| Monoclonal | |
| 0.6 μg/Test | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q8NHL6 | |
| LILRB1 | |
| Affinity chromatography | |
| RUO | |
| 10859 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG1 κ |
| Flow Cytometry | |
| HP-F1 | |
| NovaFluor Blue 610-70S | |
| LILRB1 | |
| CD85 antigen-like family member J; CD85j; Ig-like transcript 2; ILT2; ILT-2; Immunoglobulin-like transcript 2; leucocyte Ig-like receptor B1; leukocyte immunoglobulin like receptor B1; leukocyte immunoglobulin-like receptor subfamily B member 1; leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform; leukocyte immunoglobulin-like receptor subfamily B member 3-like; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LILRB1; LIR1; LIR-1; LOC100912456; MIR7; MIR-7; Monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; PIRB; PIR-B | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction