missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Mouse, PE, Clone: IT2.2, enQuireBio™
Mouse Monoclonal Antibody
183.75 € - 420.00 €
Specifications
| Antigen | CD86 |
|---|---|
| Clone | IT2.2 |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15992638
|
EnquireBio
QAB94-PE-25Tests |
25 Tests |
183.75 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15982638
|
EnquireBio
QAB94-PE-100Tests |
100 Tests |
420.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD86 | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 942 | |
| IgG2b κ | |
| Protein A/G Purified |
| IT2.2 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| NP_787058 | |
| CD86 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title