missing translation for 'onlineSavingsMsg'
Learn More

CD8a Rat, PE-Cy7, Clone: 53-6.7, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB16-PE7-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961568

  • 78.70 € / 25µg
Estimated Shipment: 20-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD8a
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd8a
Protein A/G Purified
RUO
12525
IgG2a κ
Flow Cytometry
53-6.7
PE-Cyanine7
NP_033987
Rat
25 μg
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers