missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals CD9 Immunobeads for Exosome Isolation (Mouse)

Product Code. 18305135 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 Reactions
Unit Size:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18305135 10 Reactions 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18305135 Supplier Novus Biologicals Supplier No. NBP314799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Description CD9 Immunobeads for Exosome Isolation (Mouse)
Synonym EV isolation, exosome isolation, extracellular vesicle isolation
Content And Storage Store at 4°C
Target Species Human
Quantity 10 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.