missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ M028T03B06-A
This item is not returnable.
View return policy
Description
CD90.2 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90.2 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90.2 is an 18 kDa plasma membrane antigen expressed by hematopoietic cells and neurons in all species studied, highly expressed in connective tissue, various fibroblast and stromal cell lines and lastly, by tumor endothelial cell lines. In the mouse, CD90.2 is expressed on all thymocytes and peripheral T lymphocytes.
Specifications
| CD90.2 (Thy-1.2) | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01831 | |
| Thy1 | |
| Affinity chromatography | |
| RUO | |
| 21838 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| IgG2a κ |
| Flow Cytometry | |
| 53-2.1 | |
| NovaFluor Blue 610-70S | |
| Thy1 | |
| CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction