missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (CL1040) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52961JF549
This item is not returnable.
View return policy
Description
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot
Specifications
| CD90/Thy1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| This antibody was developed using a recombinant protein derived from P04216, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL1040 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction