missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDC123 Monoclonal Antibody (24GB5705)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557359
This item is not returnable.
View return policy
Description
CDC123 Monoclonal Antibody for Western Blot, Flow
The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid. The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase, and a homodimeric dihydrolipoamide dehydrogenase. E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
Specifications
| CDC123 | |
| Monoclonal | |
| Unconjugated | |
| CDC123 | |
| AA959893; C10orf7; Cdc123; cell division cycle 123; cell division cycle 123 homolog; cell division cycle protein 123 homolog; Cell division cycle protein 123-like protein; D123; D123 gene product; G431001I09Rik; HT-1080; hypothetical protein; Protein D123; PZ32; QccE-12339 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8872 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5705 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75794 | |
| CDC123 | |
| Recombinant protein of human CDC123 (aa 1-336). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction