missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOD Recombinant Rabbit Monoclonal Antibody (JE60-47)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556541
This item is not returnable.
View return policy
Description
MYOD Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).
Specifications
| MYOD | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P15172 | |
| MYOD1 | |
| Recombinant protein within human myod1 aa 1-100/320. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| JE60-47 | |
| Unconjugated | |
| MYOD1 | |
| AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction