missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc14A Monoclonal Antibody (472508)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561442
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms.
Specifications
| Cdc14A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6GQT0, Q9UNH5 | |
| CDC14A | |
| E. coli-derived recombinant human CDC14A, Met15-Gly366. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 472508 | |
| Unconjugated | |
| CDC14A | |
| A830059A17Rik; CDC10 (cell division cycle 10, S. cerevisiae, homolog); cdc14; CDC14 cell division cycle 14 homolog A; CDC14 cell division cycle 14A; CDC14A; CDC14a1; CDC14A2; cell division cycle 14A; DFNB105; Dual specificity protein phosphatase CDC14A; hCDC14 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 229776, 310806, 8556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction