missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cdc2-Like Kinase Inhibitor, TG003

Product Code. 15456529
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456529 5 mg 5mg
15466529 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456529 Supplier Thermo Scientific Chemicals Supplier No. J64690.#0
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Cdc2-Like Kinase Inhibitor, TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members. Reports show that TG003 modifies the regulation of alternative splicing modulated by phosphorylation of SR proteins in vitro, and rescues defects in the development of Xenopus caused by an overabundance of Clk activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300801-52-9
Molecular Formula C13H15NO2S, C13H15NO2S
Synonym Clk Inhibitor; TG003

Specifications

Quantity 5 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) TG003 is a benzothiazole agent that acts as a specific and ATP-competitive inhibitor of Clk family members.
Formula Weight 249.33g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid
InChI Key BGVLELSCIHASRV-UHFFFAOYSA-N
SMILES CCN1C(SC2=C1C=C(OC)C=C2)=CC(C)=O
IUPAC Name 1-(3-ethyl-5-methoxy-2,3-dihydro-1,3-benzothiazol-2-ylidene)propan-2-one
Molecular Weight (g/mol) 249.33
Show More Show Less
compliance-icons
Product Identifier
  • Cdc2-Like Kinase Inhibitor, TG003
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (Z)-1-(3-Ethyl-5-methoxy-3H-benzothiazol-2-ylidene)propan-2-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.