missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC2L6 Antibody (CL14116) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44339
This item is not returnable.
View return policy
Description
CDC2L6 Monoclonal antibody specifically detects CDC2L6 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
Specifications
| CDC2L6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| bA346C16.3, CDC2L6, CDC2-related protein kinase 6, CDK8-like cyclin-dependent kinase, cell division cycle 2-like 6 (CDK8-like), Cell division cycle 2-like protein kinase 6, Cell division protein kinase 19, cyclin-dependent kinase (CDC2-like) 11, Cyclin-dependent kinase 11, cyclin-dependent kinase 19, Death-preventing kinase, EC 2.7.11, EC 2.7.11.22, KIAA1028CDK11 | |
| This antibody was generated using a synthetic peptide of Q9BWU1, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Protein Kinase | |
| 23097 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry | |
| CL14116 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction